Learn More
Potassium silver cyanide, 99.9% (metals basis), Ag 54.2%, Thermo Scientific Chemicals
Silver plating
101.00 € - 1320.00 €
Chemical Identifiers
CAS | 506-61-6 |
---|---|
MDL Number | MFCD00036297 |
Synonym | Potassium dicyanoargentate; Silver(I) potassium cyanide |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11346449
|
Thermo Scientific Alfa Aesar
012551.14 |
25 g |
101.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11336449
|
Thermo Scientific Alfa Aesar
012551.22 |
100 g |
318.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11356449
|
Thermo Scientific Alfa Aesar
012551.36 |
500 g |
1320.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Silver platingPotassium silver cyanide is used in silver plating and in the manufacturing of antiseptics. It is a versatile bridging ligand and finds use for multidimensional polymer construction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
506-61-6 | |
MFCD00036297 | |
Potassium dicyanoargentate; Silver(I) potassium cyanide |
Specifications
506-61-6 | |
Powder | |
MFCD00036297 | |
14,7669 | |
Soluble in waterSoluble in water; Insoluble in acids. | |
99.9% | |
Light sensitive | |
(metals basis) |
White | |
KAg(CN)2 | |
UN1588 | |
Potassium dicyanoargentate; Silver(I) potassium cyanide | |
199.01 | |
Ag 54.2% | |
2.36 g/mL | |
Potassium silver cyanide |
Safety and Handling
GHS H Statement
H300-H310-H330-H318-H290-H315
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes serious eye damage.
May be corrosive to metals.
Causes skin irritation.
P261-P262-P264b-P270-P271-P280-P301+P310-P302+P350-P304+P340-P310-P330-P361-P363-P501c
H300+H310+H330
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: CYANIDES, INORGANIC, SOLID, N.O.S.
missing translation for 'einecsNumber' : 208-047-0
missing translation for 'rtecsNumber' : TT5775000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only