Learn More
Labcold™ Powder Coated Steel Pharmacy Refrigerators
Brand:  Labcold™ RLDF18041
 View more versions of this product
Product Code. 12998604
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Microprocessor controlled temperature regulation for accuracy and stability
  • Temperature range +2°C to +8°C
  • Fan circulated interior
  • Stainless steel construction
  • Door locks fitted as standard
  • Digital temperature display with audio alarm
  • Heavy duty lockable castors on larger models
  • Optimised air circulation
Specifications
Specifications
725 x 765 x 1710mm