Learn More
Buerkle™ Sampler PowderProof™
Sampler PowderProof™ is the practically proven professional solution for mobile sampling of powders and granules.
3275.00 € - 4090.00 €
Product Code | Brand | For Use With (Application) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | Length (Metric) | Price | Quantity & Availability | ||||
10041114
|
Buerkle™
5302-0302 |
For Powders up to 2 mm dia. | 30 cm |
3275.00 €
Each |
Estimated Shipment: 16-05-2025
Log in to see stock. |
||||
10706095
|
Buerkle™
5302-1201 |
For Granules From 2mm up to 4mm dia. | 120 cm |
4090.00 €
Each |
Estimated Shipment: 16-05-2025
Log in to see stock. |
||||
11877373
|
Buerkle™
5302-0301 |
For Granules From 2 mm up to 4 mm dia. | 30 cm |
3275.00 €
Each |
Estimated Shipment: 24-07-2025
Log in to see stock. |
||||
10693585
|
Buerkle™
5302-0602 |
For Powders up to 2 mm dia. | 60 cm |
3530.00 €
Each |
In Stock
Log in to see stock. |
||||
11887373
|
Buerkle™
5302-0601 |
For Granules From 2 mm up to 4 mm dia. | 60 cm |
3530.00 €
Each |
Estimated Shipment: 16-05-2025
Log in to see stock. |
||||
11897373
|
Buerkle™
5302-0901 |
For Granules From 2 mm up to 4 mm dia. | 90 cm |
3690.00 €
Each |
In Stock
Log in to see stock. |
||||
10346964
|
Buerkle™
5302-0902 |
For Powders up to 2 mm dia. | 90 cm |
3690.00 €
Each |
Estimated Shipment: 24-07-2025
Log in to see stock. |
||||
11807383
|
Buerkle™
5302-1202 |
For Powders up to 2 mm dia. | 120 cm |
4090.00 €
Each |
Estimated Shipment: 16-05-2025
Log in to see stock. |
||||
Description
- Mobile sampling of powders and granules
- Different lances and augers can be attached to the robust drive unit and swapped without any problems
- For sampling powders and granules from drums, barrels, bags or sacks
- PowderProof® with lance for horizontal use, such as sampling from sacks, with lance for vertical use, for sampling from open containers such as barrels, drums or bags