Learn More
3M™ Versaflo™ Intrinsically Safe Powered Air Turbo TR-802E
3M™ Versaflo™ Intrinsically Safe Powered Air Turbo TR-802E is approved to Zone 0 requirements, making it suitable for areas where potentially explosive atmospheres are expected in normal operations.
Brand:  3M™ 7100169179
741.86 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 16416370
  • 1060.00 € / Each
Explore more special offers
Description
Description
  • Approved against the latest Electrical Standards
  • Easy to identify and differentiate from non-IS versions thanks to bright yellow touchpoints
  • Comfort is maximised with three user selectable flow rates and controlled airflow
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1/Cs.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
3M™ Versaflo™ Intrinsically Safe Powered Air Turbo TR-802E > Quantity: 1/Cs