missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ PowerTrack™ SYBR Green Master Mix for qPCR

PowerTrack SYBR Green Master Mix is a pre-formulated, optimized, universal 2X master mix for real-time PCR workflows with built-in visual dye–based indicators for accurate reaction set up.

97.50 € - 3235.00 €

Specifications

Content And Storage • PowerTrack SYBR Green Master Mix
• 40X Yellow Sample Buffer

Store at -20±5°C in the dark.

Detection Method SYBR
Format Tube
PCR Method qPCR
Polymerase Taq DNA Polymerase
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16505231
View Documents
Applied Biosystems™
A46012
1 mL
97.50 €
1mL
Please to purchase this item. Need a web account? Register with us today!
16525231
View Documents
Applied Biosystems™
A46109
5 mL
406.00 €
5mL
Please to purchase this item. Need a web account? Register with us today!
16535231
View Documents
Applied Biosystems™
A46110
2 x 5 mL
748.00 €
10mL
Please to purchase this item. Need a web account? Register with us today!
16545231
View Documents
Applied Biosystems™
A46111
5 x 5 mL
1728.00 €
25mL
Please to purchase this item. Need a web account? Register with us today!
16555231
View Documents
Applied Biosystems™
A46112
10 x 5 mL
3235.00 €
50mL
Please to purchase this item. Need a web account? Register with us today!
16565231
View Documents
Applied Biosystems™
A46113
50 mL
3235.00 €
50mL
Please to purchase this item. Need a web account? Register with us today!
Description
Show More
Specifications

Specifications

• PowerTrack SYBR Green Master Mix
• 40X Yellow Sample Buffer

Store at -20±5°C in the dark.

Tube
Taq DNA Polymerase
7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, QuantStudio 3, QuantStudio 5, QuantStudio 6 Flex, QuantStudio 6 Pro, QuantStudio 7 Flex, QuantStudio 7 Pro, QuantStudio 12K Flex, Bio-Rad CFX Connect™, Bio-Rad CFX384 Touch™, Bio-Rad CFX96 Touch™, BioRad CFX96™, BioRad iCycler iQ, BioRad iQ5, BioRad MyiQ, ViiA™ 7 System, StepOne™ System, MJ Chromo4, MJ Opticon, StepOnePlus™ System, Roche LightCycler 480, Stratagene Mx3000P, Stratagene Mx3005P, Stratagene Mx4000
Applied Biosystems™
Dry Ice
2X
SYBR
qPCR
Fast or Standard
ROX (Pre-mixed)
Real Time PCR SYBR Master Mix
Gene Expression, Virus Detection
DNA
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto
Applied Biosystems™ PowerTrack™ SYBR Green Master Mix for qPCR

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato