missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ PowerUp™ SYBR™ Green Master Mix for qPCR

Pre-formulated, optimized, universal 2X master mix for real-time PCR workflows.

Brand:  Applied Biosystems™ A25778

2892.75 EUR valid until 2025-12-31
Use promo code "24271" to get your promotional price.


 View more versions of this product

Product Code. 15310939

  • 3185.00 € / Pack of 10

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Coupled with user-supplied primer sets and template, PowerUp™ SYBR™ Green Master Mix is designed to amplify targets for accurate gene expression analysis.

  • A dual hot-start mechanism for excellent specificity
  • Highly reproducible CTs over a broad dynamic range
  • Inclusion of UDG to help prevent carryover contamination
  • Stability of pre-assembled reactions for up to 72 hours
  • Compatibility with most real-time qPCR instruments

Exceptional specificity with dual hot-start mechanism:
  • Specificity is paramount in obtaining high-quality data in SYBR™ Green reactions and can be enhanced through control of the Taq DNA polymerase at lower temperatures before stringent primer binding
  • PowerUp™ SYBR™ Green Master Mix uses the Dual-Lock™ Taq DNA polymerase enzyme that combines two unique hot-start mechanisms to help control its activity and to prevent undesirable early activity of the polymerase at low temperatures
  • In an experiment evaluating 24 different primer sets, PowerUp™ SYBR™ Green Master Mix generated single melt curves 100% of the time, consistent with highly specific amplification

Tight reproducibility in CTs over a wide dynamic range:
  • PowerUp™ SYBR™ Green Master Mix demonstrates excellent reproducibility over a wide dynamic range for a variety of targets tested
  • In addition, this master mix provides efficient amplification over 6 logs of sample input
  • Unlike competitor mixes that can exhibit inhibition at high cDNA inputs and low correlation coefficients over a dynamic range experiment, PowerUp™ SYBR™ Green Master Mix efficiency is maintained over the full dynamic range to help deliver maximal confidence in data quality

UDG included for carryover contamination control:
  • The inclusion of uracil-DNA glycosylase (UDG) and dUTP in the PowerUp™ SYBR™ Green Master Mix enables the degradation of any previously amplified PCR products, helping prevent contamination of subsequent qPCR reactions and possible false positives

Stability of pre-assembled reactions:
  • The tight control of the Dual-Lock™ Taq DNA polymerase in PowerUp™ SYBR™ Green Master Mix enables reactions to be assembled up to 72 hours prior to cycling without impacting data
  • This consistency allows users to have confidence and flexibility in the use of PowerUp™ SYBR™ Green Master Mix across numerous workflow scenarios1

Broad instrument compatibility:
  • PowerUp™ SYBR™ Green Master Mix can be used in either standard or fast cycling mode and is compatible with all Applied Biosystems™ real-time PCR instruments2
  • It is also compatible with the Bio-Rad IQ™5 and CFX96™/CFX384™ systems, Roche LightCycler™ LC480, and Stratagene™ MX3005P™ systems
  • For optimal results, recommended primer concentrations are 300 to 800nM

Order Info

Shipping Conditions: Wet Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

The 2X mix contains SYBR Green dye, Dual-Lock Taq DNA Polymerase, dNTPs with dUTP/dTTP blend, heat-labile UDG, ROX passive reference dye, and optimized buffer components. Contains 10 x 5 mL tube, sufficient for 5000 20-μL reactions (10 μL Master Mix per reaction).

Store at 2–8°C in the dark.
Tube
Dual-Lock Taq DNA Polymerase
7500 Fast System, 7500 System, 7900HT System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 7 Flex, StepOne™, StepOnePlus™, ViiA™ 7 System
Real Time PCR SYBR Master Mix
Wet Ice
Gene Expression
5000 Reactions
10 x 5 mL
SYBR
qPCR
Fast or Standard
PowerUP™, SYBR™
10 x 5 mL
5000 Reactions of 20 μL
2X
DNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.