missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Poxygrid™ Biohazard Bag Holders

Sturdy wire holder keeps bags open while a separate hooked ring that rests over the top keeps them securely in place

Brand:  Bel-Art™ H13187-0000

92.51 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.


 View more versions of this product

Product Code. 10287143

  • 99.40 € / Each
Estimated Shipment: 10-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Sturdy wire holder keeps bags open while a separate hooked ring that rests over the top keeps them securely in place

  • Bright biohazard orange colored epoxy-coated steel wire
  • Chemical and corrosion-resistant; unaffected by organic solvents, salts, weak acids and weak alkalis
  • Will withstand a temperature range of -223°C (-369°F) to 121°C (250°F)
  • Autoclavable
Specifications

Specifications

Biohazard Bag Holder
Autoclavable
Reusable
01-814-1, 01-814A/B, 01-815A, 01-826-5, 01-826A, 01-827B, 01-828B, 01-829B, 01-830B 24 H x 12 in. W biohazard bags
-369°F to +250°F
7.3 in.
20.5 in.
7.3 in.
Circular epoxy coated wire frame with easy-to-use hooked ring over a top
Orange
To keep biohazard bags open and ready for use
Epoxy-coated Wire
-223°C to +121°C
18.5 cm
52 cm
18.5 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bel-Art™ Poxygrid™ Biohazard Bag Holders > Accepts 12 x 24 in. bags

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.