missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Poxygrid Steel Test Tube Racks
Chemical and corrosion resistant steel test tube racks in different colors for easy identification.
Brand: Bel-Art™ SP Scienceware™ F18757-0002
This item is not returnable.
View return policy
Description
Chemical and corrosion resistant steel test tube racks in different colors for easy identification.
- Chemical and corrosion-resistant steel wire racks are coated with heat-cured epoxy
- Orange racks can be used for biohazard warning, yellow for radioactive warning
- Epoxy coatings are unaffected by organic solvents, salts, weak acids and weak alkalis
- Will withstand a temperature range of −223°C (−369°F) to 121°C (250°F)
- All racks have square holes, three decks, a slotted grid bottom and reinforced edges
Specifications
Test Tube Rack | |
Autoclavable | |
Tubes (13 to 16 mm) | |
72 | |
Non-sterile | |
-223°C to +121°C | |
16 mm |
6 x 12 | |
Yellow | |
Epoxy Coated Steel | |
Rectangle | |
13 - 16mm Tubes | |
24.2 x 13 x 6.4 cm (9.5 x 5.12 x 2.50 in.) |