missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Poxygrid Steel Test Tube Racks
Chemical and corrosion resistant steel test tube racks in different colors for easy identification.
41.95 € - 66.84 €
Specifications
Product Type | Test Tube Rack |
---|---|
Autoclavable | Autoclavable |
Temperature (Metric) Range | -223°C to +121°C |
Product Code | Brand | Array | Color | Dimensions (L x W x H) | For Use With (Equipment) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Array | Color | Dimensions (L x W x H) | For Use With (Equipment) | Price | Quantity & Availability | ||
15570012
|
Bel-Art™ SP Scienceware™
F18753-0003 |
9 x 12 | Orange | 20.5 x 16.4 x 6.4 cm (8.06 x 6.44 x 2.50 in.) | 10 - 13mm Tubes | N/A | |||
15174004
|
Bel-Art™ SP Scienceware™
F18754-0000 |
4 x 10 | Green | 20.3 x 9.2 x 6.4cm | 13 - 16mm Tubes | N/A | |||
15580012
|
Bel-Art™ SP Scienceware™
F18754-0001 |
4 x 10 | Blue | 20.3 x 9.2 x 6.4 cm (8 x 3.62 x 2.50 in.) | 13 - 16mm Tubes |
59.19 €
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
||
15510022
|
Bel-Art™ SP Scienceware™
F18755-0002 |
4 x 12 | Yellow | 24.2 x 9.2 x 6.4 cm (9.5 x 3.62 x 2.50 in.) | 13 - 16mm Tubes | N/A | |||
15540022
|
Bel-Art™ SP Scienceware™
F18757-0002 |
6 x 12 | Yellow | 24.2 x 13 x 6.4 cm (9.5 x 5.12 x 2.50 in.) | 13 - 16mm Tubes | N/A | |||
15520032
|
Bel-Art™ SP Scienceware™
F18762-0003 |
4 x 10 | Orange | 24.3 x 11 x 8.3 cm (9.56 x 4.25 x 3.25 in.) | 16 - 20mm Tubes | N/A | |||
15530032
|
Bel-Art™ SP Scienceware™
F18764-0000 |
8 x 10 | Green | 24.3 x 20.3 x 8.3 cm (9.56 x 8 x 3.25 in.) | 16 - 20mm Tubes | N/A | |||
15550032
|
Bel-Art™ SP Scienceware™
F18767-0000 |
4 x 10 | Green | 26.3 x 11.6 x 8.3 cm (10.38 x 4.56 x 3.25 in.) | 20 - 25mm Tubes |
66.84 €
Each |
Estimated Shipment: 25-02-2025
Log in to see stock. |
||
15570032
|
Bel-Art™ SP Scienceware™
F18772-0000 |
4 x 10 | Green | 29.3 x 12.8 x 8.3 cm (11.56 x 5.06 x 3.25 in.) | 20 - 25mm Tubes |
59.70 €
Each |
Estimated Shipment: 25-02-2025
Log in to see stock. |
||
15590032
|
Bel-Art™ SP Scienceware™
F18772-0002 |
4 x 10 | Yellow | 29.3 x 12.8 x 8.3 cm (11.56 x 5.06 x 3.25 in.) | 20 - 25mm Tubes | N/A | |||
15500042
|
Bel-Art™ SP Scienceware™
F18772-0003 |
4 x 10 | Orange | 29.3 x 12.8 x 8.3 cm (11.56 x 5.06 x 3.25 in.) | 20 - 25mm Tubes | N/A | |||
15510042
|
Bel-Art™ SP Scienceware™
F18773-0000 |
8 x 10 | Green | 29.3 x 24.2 x 8.3 cm (11.56 x 9.56 x 3.25 in.) | 20 - 25mm Tubes | N/A | |||
15530042
|
Bel-Art™ SP Scienceware™
F18773-0002 |
8 x 10 | Yellow | 29.3 x 24.2 x 8.3 cm (11.56 x 9.56 x 3.25 in.) | 20 - 25mm Tubes | N/A | |||
15540042
|
Bel-Art™ SP Scienceware™
F18773-0003 |
8 x 10 | Orange | 29.3 x 24.2 x 8.3 cm (11.56 x 9.56 x 3.25 in.) | 20 - 25mm Tubes |
46.90 €
Each |
Estimated Shipment: 25-02-2025
Log in to see stock. |
||
Description
Chemical and corrosion resistant steel test tube racks in different colors for easy identification.
- Chemical and corrosion-resistant steel wire racks are coated with heat-cured epoxy
- Orange racks can be used for biohazard warning, yellow for radioactive warning
- Epoxy coatings are unaffected by organic solvents, salts, weak acids and weak alkalis
- Will withstand a temperature range of −223°C (−369°F) to 121°C (250°F)
- All racks have square holes, three decks, a slotted grid bottom and reinforced edges
Specifications
Test Tube Rack | |
Autoclavable | |
-223°C to +121°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title