missing translation for 'onlineSavingsMsg'
Learn More

Melford PPO

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

Brand:  Melford 111002

 View more versions of this product

Product Code. 30017917

  • 303.00 € / 100g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

  • Substance Name: 2, 5-Diphenyloxazole
  • CAS Number: 92-71-7
  • Molecular Formula: C15H11NO
  • Molecular Weight: 221.25 g/mol
Specifications

Specifications

PPO [2, 5-Diphenyloxazole]
White
221.25
92-71-7
100 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers