missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford PPO
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
93.20 € - 1915.00 €
Specifications
Chemical Name or Material | PPO [2, 5-Diphenyloxazole] |
---|---|
CAS | 92-71-7 |
Color | White |
Molecular Weight (g/mol) | 221.25 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30017918
|
Melford
111006 |
1 kg |
1915.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30017917
|
Melford
111002 |
100 g |
303.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30017916
|
Melford
111001 |
25 g |
93.20 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
- Substance Name: 2, 5-Diphenyloxazole
- CAS Number: 92-71-7
- Molecular Formula: C15H11NO
- Molecular Weight: 221.25 g/mol
Specifications
PPO [2, 5-Diphenyloxazole] | |
92-71-7 | |
White | |
221.25 |