Learn More
Praseodymium(III) carbonate octahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
50.10 € - 430.00 €
Chemical Identifiers
CAS | 14948-62-0 |
---|---|
Molecular Formula | C3H16O17Pr2 |
Molecular Weight (g/mol) | 605.959 |
MDL Number | MFCD00211307 |
InChI Key | SEHQEMTYQGRHIC-UHFFFAOYSA-H |
Synonym | praseodymium carbonate octahydrate, 3co3.2pr.8h2o, praseodymium carbonate, praseodymium carbonate-water 2/3/8, dipraseodymium 3+ ion octahydrate trico3, praseodymium iii carbonate octahydrate, reacton, dipraseodymium 3+ octahydrate tricarbonate |
PubChem CID | 25022096 |
IUPAC Name | praseodymium(3+);tricarbonate;octahydrate |
SMILES | C(=O)([O-])[O-].C(=O)([O-])[O-].C(=O)([O-])[O-].O.O.O.O.O.O.O.O.[Pr+3].[Pr+3] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14948-62-0 | |
605.959 | |
SEHQEMTYQGRHIC-UHFFFAOYSA-H | |
25022096 | |
C(=O)([O-])[O-].C(=O)([O-])[O-].C(=O)([O-])[O-].O.O.O.O.O.O.O.O.[Pr+3].[Pr+3] |
C3H16O17Pr2 | |
MFCD00211307 | |
praseodymium carbonate octahydrate, 3co3.2pr.8h2o, praseodymium carbonate, praseodymium carbonate-water 2/3/8, dipraseodymium 3+ ion octahydrate trico3, praseodymium iii carbonate octahydrate, reacton, dipraseodymium 3+ octahydrate tricarbonate | |
praseodymium(3+);tricarbonate;octahydrate |
Specifications
14948-62-0 | |
C3H16O17Pr2 | |
praseodymium carbonate octahydrate, 3co3.2pr.8h2o, praseodymium carbonate, praseodymium carbonate-water 2/3/8, dipraseodymium 3+ ion octahydrate trico3, praseodymium iii carbonate octahydrate, reacton, dipraseodymium 3+ octahydrate tricarbonate | |
C(=O)([O-])[O-].C(=O)([O-])[O-].C(=O)([O-])[O-].O.O.O.O.O.O.O.O.[Pr+3].[Pr+3] | |
605.959 | |
605.97 (461.85 Anhydrous) | |
Reacton™ | |
(REO) |
Crystalline | |
MFCD00211307 | |
SEHQEMTYQGRHIC-UHFFFAOYSA-H | |
praseodymium(3+);tricarbonate;octahydrate | |
25022096 | |
99.99% | |
Odorless | |
Praseodymium(III) carbonate octahydrate |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 227-578-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.