Learn More
Praseodymium(III) iodide, ultra dry, 99.9+% (REO), Thermo Scientific Chemicals
132.00 € - 451.00 €
Chemical Identifiers
CAS | 13813-23-5 |
---|---|
Molecular Formula | I3Pr |
Molecular Weight (g/mol) | 521.621 |
MDL Number | MFCD00049513 |
InChI Key | PVEVRIVGNKNWML-UHFFFAOYSA-K |
Synonym | praseodymium triiodide, praseodymium iodide pri3, pri3, prazodymium iodide, praseodymium iii iodide, ultra dry |
PubChem CID | 83744 |
IUPAC Name | triiodopraseodymium |
SMILES | I[Pr](I)I |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11318168
|
Thermo Scientific Alfa Aesar
039385.03 |
1 g |
132.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11328168
|
Thermo Scientific Alfa Aesar
039385.06 |
5 g |
451.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Chemical Identifiers
13813-23-5 | |
521.621 | |
PVEVRIVGNKNWML-UHFFFAOYSA-K | |
83744 | |
I[Pr](I)I |
I3Pr | |
MFCD00049513 | |
praseodymium triiodide, praseodymium iodide pri3, pri3, prazodymium iodide, praseodymium iii iodide, ultra dry | |
triiodopraseodymium |
Specifications
737°C | |
Crystals or Needles | |
PrI3 | |
praseodymium triiodide, praseodymium iodide pri3, pri3, prazodymium iodide, praseodymium iii iodide, ultra dry | |
PVEVRIVGNKNWML-UHFFFAOYSA-K | |
triiodopraseodymium | |
83744 | |
≥99.9% | |
Hygroscopic; Light sensitive | |
Praseodymium(III) iodide, Ultra dry |
13813-23-5 | |
I3Pr | |
MFCD00049513 | |
Soluble in water. | |
I[Pr](I)I | |
521.621 | |
521.62 | |
Ampule | |
(REO) |
Safety and Handling
GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
H317-H334
missing translation for 'einecsNumber' : 237-466-1
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
- Ampouled under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only