Learn More
Fisherbrand™ Precision Balances
Featuring RS232 connectivity for easy communication, a backlit display and a simple interface for uncomplicated operation, these balances are perfectly designed for your workplace.
1065.00 € - 1675.00 €
Specifications
Certifications/Compliance | GLP with optional printer compliance |
---|---|
Connection Type | RS232 |
Display Type | Backlit LCD |
Includes | AC adapter |
Warranty | 2 Years |
Product Code | Brand | Capacity | Model | Calibration | Dimensions (D x W x H) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Model | Calibration | Dimensions (D x W x H) | Price | Quantity & Availability | ||
15947500
|
Fisherbrand™
30488139 |
220 g | FPRS223 | Internal | 317 x 201 x 303 mm |
1100.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15937500
|
Fisherbrand™
30488138 |
220 g | FPRS223/E | External | 317 x 201 x 303 mm |
1675.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15967500
|
Fisherbrand™
30488141 |
420 g | FPRS423 | Internal | 317 x 201 x 303 mm |
1200.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15957500
|
Fisherbrand™
30488140 |
420 g | FPRS423/E | External | 317 x 201 x 303 mm |
1130.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15987500
|
Fisherbrand™
30488143 |
820 g | FPRS822 | Internal | 317 x 201 x 93 mm |
1065.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15977500
|
Fisherbrand™
30488142 |
820 g | FPRS822/E | External | 317 x 201 x 93 mm |
1170.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15907510
|
Fisherbrand™
30488145 |
2200 g | FPRS2202 | Internal | 317 x 201 x 93 mm |
1150.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15997500
|
Fisherbrand™
30488144 |
2200 g | FPRS2202/E | External | 317 x 201 x 93 mm |
1085.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15947510
|
Fisherbrand™
30488150 |
4200 g | FPRS4201 | Internal | 317 x 201 x 93 mm |
1075.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15937510
|
Fisherbrand™
30488149 |
4200 g | FPRS4201/E | External | 317 x 201 x 93 mm |
1220.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15927510
|
Fisherbrand™
30488147 |
4200 g | FPRS4202 | Internal | 317 x 201 x 93 mm |
1250.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15917510
|
Fisherbrand™
30488146 |
4200 g | FPRS4202/E | External | 317 x 201 x 93 mm |
1120.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15957510
|
Fisherbrand™
30488151 |
6200 g | FPRS6201/E | External | 317 x 201 x 93 mm |
1120.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Standard RS-232 connectivity for easy data transfer
- Stainless steel platform
- Two adjustable feet that allow user to level the balance
- Up-front level indicator - no need to look in back of balance during leveling process
- Smart design and durable construction - small footprint which saves on benchtop space while providing a large weighing surface
- Large, bright backlit display allows for easy viewing of results
- Four mechanical keys enable easy operation of basic application modes
- Weighing modes include weighing, parts counting and percent weighing
- Includes AC adapter
- Certifications: FCC, CE
- Two year warranty
Specifications
GLP with optional printer compliance | |
Backlit LCD | |
2 Years | |
100/240 V ∼200 mA 50/60 Hz 12/18 VA | |
50/60 Hz |
RS232 | |
AC adapter | |
Weighing, Parts Counting, Percent Weighing | |
0.5 A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ Precision Balances