Learn More
JULABO PRESTO Process Systems
GREENER_CHOICE
The PRESTO systems are designed for precise temperature control as well as rapid temperature changes, making them ideal for reactor vessels, material stress tests, or temperature simulations.
Brand:  JULABO 9420381.N1
 View more versions of this product
Product Code. 17714869
  • 26835.00 € / Each
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Wide working temperature ranges without changing fluids
  • Ethernet interface
  • SD-Card slot
  • pace optimized design creates more room directly next to the units
  • Connections for USB, Ethernet, RS232, and Alarm output
Specifications
Specifications
-30°C to +250°C
29.5 x 13 x 26.4 in.
A38
Process System