missing translation for 'onlineSavingsMsg'
Learn More

Tuttnauer Printer Paper Roll

Printer Paper Roll for SRV000-0485.

Brand:  Tuttnauer THE002-0118

Product Code. 17849598

  • 16.30 € / Each
Estimated Shipment: 21-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Thermal paper 57 mm width
  • Roll for printer model TDP-225
Specifications

Specifications

Printer SRV000-0485
Printer Paper
5.1 in.
57 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tuttnauer Printer Paper Roll > Printer Paper Roll

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.