Learn More
Epredia™ PrintMate™ Biopsy Cassettes
Epredia™ PrintMate Cassettes™ are designed to contain incised tissue safely and securely for tissue processing, embedding and sectioning. PrintMate Cassettes provide exceptional scanning accuracy and reliability when used with the Epredia™ PrintMate™ and Epredia™ SlideMate™ barcode scanner.
Brand:  Epredia™ B851080LG
 View more versions of this product
Product Code. 16353986
  • 138.00 € / Pack of 1000
Explore more special offers
Description
Description
Designed for performance and user convenience, the PrintMate cassette range has been optimized for superior workflow and printing with the PrintMate Classic and PrintMate AS printing systems. The even, textured printing face provides the optimal surface for high-quality printing every time. For additional information, the right and left sides are textured for clear handwriting with suitable pens. The lid catches have been adjusted for ease of use with low force opening and closing, while still giving the security needed while processing.Available in tubes for automatic loading or loose for manual use, and in twelve colors for enhanced workflow identification.
Specifications
Specifications
Small Square Pores
Four Dispensers of 250 Cassettes or 1000 per Case.
Yes
PrintMate Biopsy Cassette for PrintMate Printers
Separate lids included
Reversible
Light Green