Learn More
Epredia™ PrintMate™ Biopsy Cassettes
Epredia™ PrintMate Cassettes™ are designed to contain incised tissue safely and securely for tissue processing, embedding and sectioning. PrintMate Cassettes provide exceptional scanning accuracy and reliability when used with the Epredia™ PrintMate™ and Epredia™ SlideMate™ barcode scanner.
39.85 € - 146.00 €
Specifications
PrintMate Compatible Yes
View More Specs
Products 34
Product Code Brand Color Quantity Price Quantity & Availability  
Product Code Brand Color Quantity Price Quantity & Availability  
15487604
View Documents
Epredia™
A84210050
Aqua N/A N/A N/A
16313986
View Documents
Epredia™
B851080BE
Beige 4 x 250 cassettes (1000)/cs
138.00 €
Pack of 1000
15497604
View Documents
Epredia™
A84210051
Blue N/A N/A N/A
16232321
View Documents
Epredia™
B851080BL
Blue 4 x 250 cassettes (1000)/cs
146.00 €
Pack of 1000
15407614
View Documents
Epredia™
A84210052
Green N/A N/A N/A
16323986
View Documents
Epredia™
B851080GN
Green 4 x 250 cassettes (1000)/cs
138.00 €
Pack of 1000
15417614
View Documents
Epredia™
A84210053
Gray N/A N/A N/A
16333986
View Documents
Epredia™
B851080GY
Gray 4 x 250 cassettes (1000)/cs
138.00 €
Pack of 1000
15427614
View Documents
Epredia™
A84210054
Lilac N/A N/A N/A
16343986
View Documents
Epredia™
B851080LA
Lavender 4 x 250 cassettes (1000)/cs
138.00 €
Pack of 1000
15437614
View Documents
Epredia™
A84210055
Orange N/A N/A N/A
16353986
View Documents
Epredia™
B851080LG
Light Green 4 x 250 cassettes (1000)/cs
138.00 €
Pack of 1000
15447614
View Documents
Epredia™
A84210056
Pink N/A N/A N/A
16363986
View Documents
Epredia™
B851080LI
Lilac 4 x 250 cassettes (1000)/cs
138.00 €
Pack of 1000
15457614
View Documents
Epredia™
A84210057
Tan N/A N/A N/A
16373986
View Documents
Epredia™
B851080OR
Orange 4 x 250 cassettes (1000)/cs
138.00 €
Pack of 1000
15467614
View Documents
Epredia™
A84210058
White N/A N/A N/A
16383986
View Documents
Epredia™
B851080PK
Pink 4 x 250 cassettes (1000)/cs
138.00 €
Pack of 1000
15477614
View Documents
Epredia™
A84210059
Yellow N/A N/A N/A
16393986
View Documents
Epredia™
B851080TN
Tan 4 x 250 cassettes (1000)/cs
138.00 €
Pack of 1000
16303996
View Documents
Epredia™
B851080WH
White 4 x 250 cassettes (1000)/cs
138.00 €
Pack of 1000
16313996
View Documents
Epredia™
B851080YW
Yellow 4 x 250 cassettes (1000)/cs
138.00 €
Pack of 1000
16354016
View Documents
Epredia™
B851110BE
Beige 10 x 75 cassettes (750)/cs
113.00 €
Pack of 750
16364016
View Documents
Epredia™
B851110BL
Blue 10 x 75 cassettes (750)/cs
113.00 €
Pack of 750
16374016
View Documents
Epredia™
B851110GN
Green 10 x 75 cassettes (750)/cs
113.00 €
Pack of 750
16384016
View Documents
Epredia™
B851110GY
Gray 10 x 75 cassettes (750)/cs
113.00 €
Pack of 750
16394016
View Documents
Epredia™
B851110LA
Lavender 10 x 75 cassettes (750)/cs
113.00 €
Pack of 750
16304026
View Documents
Epredia™
B851110LG
Light Green 10 x 75 cassettes (750)/cs
113.00 €
Pack of 750
16314026
View Documents
Epredia™
B851110LI
Lilac 10 x 75 cassettes (750)/cs
113.00 €
Pack of 750
16324026
View Documents
Epredia™
B851110OR
Orange 10 x 75 cassettes (750)/cs
113.00 €
Pack of 750
16334026
View Documents
Epredia™
B851110PK
Pink 10 x 75 cassettes (750)/cs
113.00 €
Pack of 750
16344026
View Documents
Epredia™
B851110TN
Tan 10 x 75 cassettes (750)/cs
113.00 €
Pack of 750
16354026
View Documents
Epredia™
B851110WH
White 10 x 75 cassettes (750)/cs
113.00 €
Pack of 750
16364026
View Documents
Epredia™
B851110YW
Yellow 10 x 75 cassettes (750)/cs
113.00 €
Pack of 750
Description
Description
Designed for performance and user convenience, the PrintMate cassette range has been optimized for superior workflow and printing with the PrintMate Classic and PrintMate AS printing systems.  The even, textured printing face provides the optimal surface for high-quality printing every time.  For additional information, the right and left sides are textured for clear handwriting with suitable pens.  The lid catches have been adjusted for ease of use with low force opening and closing, while still giving the security needed while processing.
Available in tubes for automatic loading or loose for manual use, and in twelve colors for enhanced workflow identification.
Specifications
Specifications
Yes
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Epredia™ PrintMate™ Biopsy Cassettes