missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Techsit™ Pro-Tech Laboratory High Chair
Advanced laboratory high chair is fully adjustable with an adjustable foot ring.
728.92 € - 857.18 €
Specifications
Adjustable | Yes |
---|---|
Back Style | Ergonomic |
Finish | Vinyl |
Frame Material | Steel |
Lift Type | Gas Lift |
Product Code | Brand | Color | With Armrest | With Casters | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | With Armrest | With Casters | Price | Quantity & Availability | |||
16503914
|
Techsit™
TS7002B/BL |
Black | No | Yes |
775.24 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
16513914
|
Techsit™
TS7002G |
Gray | No | No |
728.92 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
16523914
|
Techsit™
TS7002G/AA |
Gray | Yes | No |
857.18 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
16533914
|
Techsit™
TS7002G/BL |
Gray | No | Yes |
775.24 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
16583904
|
Techsit™
TS7002B |
Black | No | No |
728.92 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
16593904
|
Techsit™
TS7002B/AA |
Black | Yes | Yes |
857.18 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
Description
- Vinyl high chair with adjustable foot ring
- Fully adjustable seat height, back height, and seat depth
- Ergonomically designed chair that automatically adjusts to the user
- Cleanroom Class 7 suitable
Specifications
Yes | |
Vinyl | |
Gas Lift | |
High Bench Height | |
5 Year | |
Yes | |
Yes |
Ergonomic | |
Steel | |
150 kg | |
Vinyl | |
Yes | |
Yes | |
Laboratory Seat |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title