missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Contec™ ProChlor
Contec ProChlor is a revolutionary new sporicide achieving a 100% kill against spores in under 1 min.
Marke: Contec™ SBT102PC
227.99 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Ensures cleanroom is free from contamination and particulates.
A blend of hypochlorous acid in purified water, Contec ProChlor is provided ready-to-use and is efficacious against bacteria, fungi, moulds, yeasts, viruses and spores.
Contec ProChlor is 0.2 micron filtered, and filled in a Grade A environment. Provided double bagged, the product is designed for ease of entry into pharmaceutical cleanrooms.
Features & Benefits
- Log 6 kill against Bacillus subtilis in 60 secs saves time spent on biodecontamination
- Filtered to 0.2 microns in a Grade B cleanroom ensures the product is free from contamination and particulates
- Sterile version available for Grade A and B cleanrooms
- Contains no quaternary ammonium or surfactant so it's very low residue
- No hazard classification means only basic PPE required and no special disposal is required
- Double bagged packed in linear tear packaging so it's easy to open even when wearing gloves
- Not classed as corrosive and can be used safely in all areas of the cleanroom
Spezifikation
Sporicide | |
3 to 6 | |
Colorless | |
Clear | |
2000 ppm hypochlorous acid | |
Do not freeze. Store below 40°C. | |
Filtered | |
Ready-to-Use Hard Surface Disinfectant | |
6 x 1 L | |
1 L |
Liquid Spray | |
Sterile | |
Not Autoclavable | |
Stabilized Hypochlorous acid blended with purified water | |
Trigger Spray Bottle | |
Limited-use | |
No | |
Double bagged in polyethylene | |
33.81 oz. |