missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Contec™ ProChlor
Contec ProChlor is a revolutionary new sporicide achieving a 100% kill against spores in under 1 min.
148.00 € - 472.00 €
Specifications
Appearance | Colorless |
---|---|
Autoclavable | Not Autoclavable |
Color | Clear |
Filtered | Filtered |
Flammable | No |
Product Code | Brand | Container Type | Quantity | Sterility | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Container Type | Quantity | Sterility | Price | Quantity & Availability | |||
15547171
|
Contec™
SBT102PC |
Trigger Spray Bottle | 6 x 1 L | Sterile |
472.00 €
6 bottles |
In Stock
Log in to see stock. |
|||
15567161
|
Contec™
FBT102PC |
Trigger Spray Bottle | 6 x 1 L | Non-sterile |
148.00 €
6 bottles |
In Stock
Log in to see stock. |
|||
15597161
|
Contec™
SBC502PC |
Capped Bottle | 2 x 5 L | Sterile |
291.00 €
2 bottles |
In Stock
Log in to see stock. |
|||
15904332
|
Contec™
SBC102PC |
Capped Bottle | 6 x 1 L | Sterile |
277.00 €
6 bottles |
Estimated Shipment: 06-01-2025
Log in to see stock. |
|||
Description
Ensures cleanroom is free from contamination and particulates.
A blend of hypochlorous acid in purified water, Contec ProChlor is provided ready-to-use and is efficacious against bacteria, fungi, moulds, yeasts, viruses and spores.
Contec ProChlor is 0.2 micron filtered, and filled in a Grade A environment. Provided double bagged, the product is designed for ease of entry into pharmaceutical cleanrooms.
Features & Benefits
- Log 6 kill against Bacillus subtilis in 60 secs saves time spent on biodecontamination
- Filtered to 0.2 microns in a Grade B cleanroom ensures the product is free from contamination and particulates
- Sterile version available for Grade A and B cleanrooms
- Contains no quaternary ammonium or surfactant so it's very low residue
- No hazard classification means only basic PPE required and no special disposal is required
- Double bagged packed in linear tear packaging so it's easy to open even when wearing gloves
- Not classed as corrosive and can be used safely in all areas of the cleanroom
Specifications
Colorless | |
Clear | |
No |
Not Autoclavable | |
Filtered |
Safety and Handling
missing translation for 'shelfLife' : 2 Years