missing translation for 'onlineSavingsMsg'
Learn More

Propargyl alcohol, 99%, Thermo Scientific Chemicals

24.82 € - 197.99 €

Chemical Identifiers

CAS 107-19-7
Molecular Formula C3H4O
Molecular Weight (g/mol) 56.06
MDL Number MFCD00002912
InChI Key TVDSBUOJIPERQY-UHFFFAOYSA-N
Synonym propargyl alcohol, 2-propyn-1-ol, ethynylcarbinol, 1-propyn-3-ol, propynyl alcohol, 2-propynyl alcohol, 2-propynol, 3-propynol, 1-hydroxy-2-propyne, 3-hydroxy-1-propyne
PubChem CID 7859
ChEBI CHEBI:28905
IUPAC Name prop-2-yn-1-ol
SMILES OCC#C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11474033
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A10295.AE
100 mL
24.82 €
100mL
Please to purchase this item. Need a web account? Register with us today!
11484033
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A10295.AP
500 mL
50.65 €
500mL
Please to purchase this item. Need a web account? Register with us today!
11494033
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A10295.0F
2500 mL
197.99 €
2500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Propargyl alcohol is a solvent stabilizer that is useful as an intermediate in organic synthesis, an electroplating brightener additive. It is useful 3-carbon fragment in acetylene coupling reactions, It undergoes Pd-catalyzed O-coupling with alcohols to give the corresponding 1-methoxyallyl ethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propargyl alcohol is a solvent stabilizer that is useful as an intermediate in organic synthesis, an electroplating brightener additive. It is useful 3-carbon fragment in acetylene coupling reactions, It undergoes Pd-catalyzed O-coupling with alcohols to give the corresponding 1-methoxyallyl ethers.

Solubility
Fully miscible in water.

Notes
Store at 4°C. Incompatible with oxidizing agent, strong acids, strong bases, amines, phosphorous pentoxide, isocyanate vapor & heat.
Specifications

Chemical Identifiers

107-19-7
56.06
TVDSBUOJIPERQY-UHFFFAOYSA-N
7859
prop-2-yn-1-ol
C3H4O
MFCD00002912
propargyl alcohol, 2-propyn-1-ol, ethynylcarbinol, 1-propyn-3-ol, propynyl alcohol, 2-propynyl alcohol, 2-propynol, 3-propynol, 1-hydroxy-2-propyne, 3-hydroxy-1-propyne
CHEBI:28905
OCC#C

Specifications

107-19-7
0.949
33°C (91°F)
HC≡CCH2OH
MFCD00002912
506003
propargyl alcohol, 2-propyn-1-ol, ethynylcarbinol, 1-propyn-3-ol, propynyl alcohol, 2-propynyl alcohol, 2-propynol, 3-propynol, 1-hydroxy-2-propyne, 3-hydroxy-1-propyne
TVDSBUOJIPERQY-UHFFFAOYSA-N
prop-2-yn-1-ol
7859
56.06
Propargyl alcohol
-53°C
114°C to 115°C
C3H4O
1.432
UN2929
14,7809
Fully miscible in water.
OCC#C
56.06
CHEBI:28905
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H314-H226
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H226-H301-H310+H330-H314-H335-H373

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 203-471-2

missing translation for 'rtecsNumber' : UK5075000

missing translation for 'tsca' : Yes

Recommended Storage : Keep cold

Product Identifier
  • Propargyl alcohol

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H310-Fatal in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : 2-Propyn-1-ol

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only