missing translation for 'onlineSavingsMsg'
Learn More
Propargyl alcohol, 99%, Thermo Scientific Chemicals Product Code.: 11484033

Propargyl alcohol, 99%, Thermo Scientific Chemicals

Product Code. 11484033
500 mL
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
This item is not returnable. View return policy

Product Code. 11484033

Brand: Thermo Scientific Alfa Aesar A10295.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Propargyl alcohol is a solvent stabilizer that is useful as an intermediate in organic synthesis, an electroplating brightener additive. It is useful 3-carbon fragment in acetylene coupling reactions, It undergoes Pd-catalyzed O-coupling with alcohols to give the corresponding 1-methoxyallyl ethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propargyl alcohol is a solvent stabilizer that is useful as an intermediate in organic synthesis, an electroplating brightener additive. It is useful 3-carbon fragment in acetylene coupling reactions, It undergoes Pd-catalyzed O-coupling with alcohols to give the corresponding 1-methoxyallyl ethers.

Solubility
Fully miscible in water.

Notes
Store at 4°C. Incompatible with oxidizing agent, strong acids, strong bases, amines, phosphorous pentoxide, isocyanate vapor & heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-19-7
Molecular Formula C3H4O
Molecular Weight (g/mol) 56.06
MDL Number MFCD00002912
InChI Key TVDSBUOJIPERQY-UHFFFAOYSA-N
Synonym propargyl alcohol, 2-propyn-1-ol, ethynylcarbinol, 1-propyn-3-ol, propynyl alcohol, 2-propynyl alcohol, 2-propynol, 3-propynol, 1-hydroxy-2-propyne, 3-hydroxy-1-propyneShow MoreShow Less
PubChem CID 7859
ChEBI CHEBI:28905
IUPAC Name prop-2-yn-1-ol
SMILES OCC#C

Specifications

Melting Point -53°C
Density 0.949
Boiling Point 114°C to 115°C
Flash Point 33°C (91°F)
Linear Formula HC≡CCH2OH
Refractive Index 1.432
Quantity 500 mL
UN Number UN2929
Beilstein 506003
Merck Index 14,7809
Solubility Information Fully miscible in water.
Formula Weight 56.06
Percent Purity 99%
Chemical Name or Material Propargyl alcohol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10238224 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA10295.0F,A10295.AE,A10295.AP
Lot NumberY17H013 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA10295.0F,A10295.AE,A10295.AP
Lot Number10133712 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10295.0F,A10295.AE,A10295.AP
Lot Number10163478 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10295.0F,A10295.AE,A10295.AP
Lot Number61200047 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10295.0F,A10295.AE,A10295.AP

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Propargyl alcohol, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.