missing translation for 'onlineSavingsMsg'
Learn More

Propargyl bromide, 97%, 80% w/w in toluene, stab. with magnesium oxide, Thermo Scientific Chemicals

Product Code. 11322886
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Propargyl bromide is used as an alkylating agent as well as an intermediate for the synthesis of organic compounds, agrochemicals and pharmaceuticals. It reacts with magnesium in presence of ether to get a Grignard reagent namely allenylmagnesium bromide, which on further reaction with propargyl bromide prepares bis-allene 1,2,4,5-hexatetraene. Also, used as metal brightening agents, corrosion inhibitor and soil fumigant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propargyl bromide is used as an alkylating agent as well as an intermediate for the synthesis of organic compounds, agrochemicals and pharmaceuticals. It reacts with magnesium in presence of ether to get a Grignard reagent namely allenylmagnesium bromide, which on further reaction with propargyl bromide prepares bis-allene 1,2,4,5-hexatetraene. Also, used as metal brightening agents, corrosion inhibitor and soil fumigant.

Solubility
Miscible with ethanol, ether, benzene, carbon tetrachloride and chloroform. Immiscible with water.

Notes
Light sensitive. Store in a cool place. Incompatible withbases, strong oxidizing agents, copper, peroxides, silver, silver oxides, mercury and mercury oxides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-96-7
Molecular Formula C3H3Br
Molecular Weight (g/mol) 118.961
MDL Number MFCD00000241
InChI Key YORCIIVHUBAYBQ-UHFFFAOYSA-N
Synonym propargyl bromide, 3-bromopropyne, 3-bromo-1-propyne, 1-propyne, 3-bromo, 2-propynyl bromide, propynyl bromide, 1-bromo-2-propyne, propyne, 3-bromo, gamma-bromoallylene, 1-brom-2-propin
PubChem CID 7842
IUPAC Name 3-bromoprop-1-yne
SMILES C#CCBr

Specifications

Melting Point -61°C
Density 1.335
Boiling Point 88°C to 90°C
Flash Point 3°C (37°F)
Linear Formula HC≡CCH2Br
Refractive Index 1.494
Quantity 100 g
UN Number UN2924
Beilstein 605309
Sensitivity Light sensitive
Solubility Information Miscible with ethanol,ether,benzene,carbon tetrachloride and chloroform. Immiscible with water.
Formula Weight 118.97
Percent Purity 97%
Concentration or Composition (by Analyte or Components) 80% w/w in toluene
Chemical Name or Material Propargyl bromide, stabilized with magnesium oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.