Learn More
Contec™ PROSAT™ Sterile Presaturated Nonwoven Wipes
Wipe down isolators, cabinets, hoods, and other cleanroom equipment and tools. Contec™ PROSAT™ Sterile Presaturated Nonwoven Wipes are ideal for cleanup following BD RODAC™ plate sampling. These wipes are designed specifically for sterile environments.
16.20 € - 1055.00 €
Product Code | Brand | Dimensions (L x W) | Packaging Quantity | Saturation Solution | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Packaging Quantity | Saturation Solution | Price | Quantity & Availability | |||
17224513
|
Contec™
PS-7030IR-BPR |
22.86 x 22.86 cm (9 x 9 in.) | 20 Pk. | 70% IPA, 30% DI water |
895.00 €
Pack of 800 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17274513
|
Contec™
PS-911EB-BPR |
23 x 28 cm (9 x 11 in.) | 30 Pk. | 70% IPA, 30% DI water |
965.00 €
Pack of 1440 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17224523
|
Contec™
PSC20001-BPR |
23 x 28 cm (9 x 11 in.) | 24 Pk. | 70% IPA, 30% DI water |
1055.00 €
Pack of 1152 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17244523
|
Contec™
PSC20005-BPR |
23 x 28 cm (9 x 11 in.) | 50 Pk. | 70% USP grade IPA, 30% DI water |
885.00 €
Pack of 1400 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17264523
|
Contec™
PSCP0001-BPR |
15 x 23 cm (6 x 9 in.) | 50 Pk. | 70% USP grade IPA, 30% DI water | |||||
11917025
|
Contec™
PSC20001 |
27.9 x 23 cm (11 x 9 in.) | 24 Pk. | 70% IPA, 30% DI Water | |||||
16294072
|
Contec™
PS-7030IR |
22.86 x 22.86 cm (9 x 9 in.) | 40 Packs of 20 Wipes | 70% IPA, 30% DI Water | |||||
Description
Wipe down isolators, cabinets, hoods, and other cleanroom equipment and tools. Contec™ PROSAT™ Sterile Presaturated Nonwoven Wipes are ideal for cleanup following BD RODAC™ plate sampling. These wipes are designed specifically for sterile environments.
- Designed for pharmaceutical, biotech, and medical device applications
- Choose spunlaced nonwoven 100% polyester wipes for improved cleanliness, economical hydroentangled polyester/cellulose wipers, or meltblown polypropylene wipes for improved durability
- Conveniently presaturated with 70/30 blend of semi-conductor grade IPA and DI water to reduce VOC emissions and hazardous waste
- All wipes are sterilized by gamma radiation to a 10-6 Sterility Assurance Level and validated per AAMI ISO 11137 (Method 1) guidelines
- Certificate of Sterility and Certificate of Irradiation included
- Environment: ISO 5-8, Grade A/B
- Individually lot numbered resealable pouches with tamper-evident labels ensure product integrity and cleanliness
Compliance
Validated sterile to a Sterility Assurance Level of 10-6 in accordance with AAMI/ISO 1137 Method 1 Guidelines.
Specifications
Pre-saturated Wipe | |
Sterile |
Safety and Handling
- CONTEC PROSAT STERILE AMP DELTA 70/30 IPA & DI WATER
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use dry chemical, CO2 to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
- MIXTURE LIST-Contains : propan-2-ol