Learn More
Contec™ PROSAT™ Sterile Presaturated Nonwoven Wipes

Wipe down isolators, cabinets, hoods, and other cleanroom equipment and tools. Contec™ PROSAT™ Sterile Presaturated Nonwoven Wipes are ideal for cleanup following BD RODAC™ plate sampling. These wipes are designed specifically for sterile environments.
Brand: Contec™ PS-911EB-BPR
793.66 EUR valid until 2025-05-02
Use promo code "24151" to get your promotional price.
Description
Wipe down isolators, cabinets, hoods, and other cleanroom equipment and tools. Contec™ PROSAT™ Sterile Presaturated Nonwoven Wipes are ideal for cleanup following BD RODAC™ plate sampling. These wipes are designed specifically for sterile environments.
- Designed for pharmaceutical, biotech, and medical device applications
- Choose spunlaced nonwoven 100% polyester wipes for improved cleanliness, economical hydroentangled polyester/cellulose wipers, or meltblown polypropylene wipes for improved durability
- Conveniently presaturated with 70/30 blend of semi-conductor grade IPA and DI water to reduce VOC emissions and hazardous waste
- All wipes are sterilized by gamma radiation to a 10-6 Sterility Assurance Level and validated per AAMI ISO 11137 (Method 1) guidelines
- Certificate of Sterility and Certificate of Irradiation included
- Recommended for use in ISO Class 5 (FED-STD-209E Class 100/M3.5) environments
- Individually lot numbered resealable pouches with tamper-evident labels ensure product integrity and cleanliness
Compliance
Validated sterile to a Sterility Assurance Level of 10-6 in accordance with AAMI/ISO 1137 Method 1 Guidelines.

Specifications
Pre-saturated Wipe | |
23 cm | |
27.9 cm | |
ISO Class 4 | |
Ideal for Routine Cleaning and Wipe Down of Lab Tools, Instruments and Other Equipment and Designed for Sterile Environments | |
70% IPA, 30% DI water |
9 in. | |
11 in. | |
Polypropylene | |
23 x 28 cm (9 x 11 in.) | |
30 Pk. | |
Sterile |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- CONTEC PROSAT MBPP WITH 70% IPA
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use dry chemical, carbondioxide to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
- MIXTURE LIST-Contains : propan-2-ol
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1111664-0-0-1 | Certificate of Quality | 03/04/2025 | PS-911EB-BPR |
1095943-0-0-1 | Certificate of Quality | 06/03/2025 | PS-911EB-BPR |
1095942-0-0-1 | Certificate of Quality | 01/03/2025 | PS-911EB-BPR |
1095940-0-0-1 | Certificate of Quality | 17/01/2025 | PS-911EB-BPR |
1078779-0-0-1 | Certificate of Quality | 14/01/2025 | PS-911EB-BPR |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.