Learn More
Laboratoires Anios™ Protective Cream Bottle
Protective cream for hands. Glycerin and shea butter present in this protective cream bring the essential ingredients to protect hands irritated by the stresses and strains of daily life, its concentrated, non-greasy formula penetrates and acts rapidly.
Brand: Laboratoires Anios™ 399236DK
Description
White Cream - Honeysuckle Perfume
- ph at 20° : 6,5-7,5
- viscosity : 120 000cp - 280 000 cp
Specifications
Protective Hand Cream | |
Bottle | |
500 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.