missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Protein LoBind™ Deepwell Plates
BIOPHARMAGREENER_CHOICE

Eppendorf Protein LoBind Plates are designed for use in proteomics or other fields of protein research, where protein concentration tend to be very small and sample recovery is vital for assay results.

Brand:  Eppendorf™ 0030504305

408.50 EUR valid until 2024-12-31
Use promo code "21693" to get your promotional price.


 View more versions of this product

Product Code. 11518643

  • 441.76 € / Pack of 20
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Eppendorf LoBind material ensures excellent sample recovery for improved assay results.
  • Free of surface coating (e.g., silicone) to minimize the risk of sample interference.
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors.
  • High-contrast Unique OptiTrack™ matrix: up to 30% faster sample identification and fewer pipetting errors.
  • RecoverMax™ well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties.
  • Raised well rims and a smooth surface ensure reliable sealing in plates.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Standard
Autoclavable
White
Polypropylene
96
Non-sterile
Proteomics or other fields of protein research
20 Pk.
Yes
85.5 x 127.8 x 60 mm
8 x 12
20 Plates (5 Bags x 4 Plates)
Protein LoBind™ Plate
2000 μL
50 to 2,000 μL
Yes
6000 x g
20 Pk.
Colorless
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.