Learn More
Eppendorf™ Protein LoBind™ Deepwell Plates
BIOPHARMAGREENER_CHOICE
Eppendorf Protein LoBind Plates are designed for use in proteomics or other fields of protein research, where protein concentration tend to be very small and sample recovery is vital for assay results.
375.50 € - 2115.00 €
Specifications
Autoclavable Autoclavable
Material Polypropylene
Sterility Non-sterile
Max. RCF 6000 x g
Well Color Colorless
View More Specs
Products 5
Product Code Brand Color No. of Wells Volume (Metric) Well Packaging Price Quantity & Availability  
Product Code Brand Color No. of Wells Volume (Metric) Well Packaging Price Quantity & Availability  
10574073
BIOPHARMAGREENER_CHOICE
View Documents
Eppendorf™
0030504119
Yellow 96 500 μL 5 Bags x 8 Plates
784.00 €
Pack of 40
Estimated Shipment: 04-04-2025
to see stock.
Add to basket
10367133
BIOPHARMAGREENER_CHOICE
View Documents
Eppendorf™
0030508106
White 96 500 μL 10 Bags x 12 Plates
2115.00 €
Pack of 120
In Stock
to see stock.
Add to basket
10709263
BIOPHARMAGREENER_CHOICE
View Documents
Eppendorf™
0030504208
White 96 1000 μL 5 Bags x 4 Plates
375.50 €
Pack of 20
In Stock
to see stock.
Edge
Add to basket
10215373
BIOPHARMAGREENER_CHOICE
View Documents
Eppendorf™
0030508203
White, Clear Bottom 96 1000 μL 10 Bags x 8 Plates
1295.00 €
Pack of 80
In Stock
to see stock.
Add to basket
11518643
BIOPHARMAGREENER_CHOICE
View Documents
Eppendorf™
0030504305
White 96 2000 μL 5 Bags x 4 Plates
441.76 €
Pack of 20
In Stock
to see stock.
Add to basket
Description
Description
  • Eppendorf LoBind material ensures excellent sample recovery for improved assay results.
  • Free of surface coating (e.g., silicone) to minimize the risk of sample interference.
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors.
  • High-contrast Unique OptiTrack™ matrix: up to 30% faster sample identification and fewer pipetting errors.
  • RecoverMax™ well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties.
  • Raised well rims and a smooth surface ensure reliable sealing in plates.
Specifications
Specifications
Autoclavable
Non-sterile
Colorless
Polypropylene
6000 x g