missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Protein LoBind™ Tubes

Engineered to minimize sample binding and elevate recovery, Eppendorf™ Protein LoBind™ Tubes safeguard your valuable proteins during processing. With cutting-edge LoBind technology, ensure optimal yields for your Western blotting, mass spectrometry, and protein purification applications.

Brand:  Eppendorf™ 0030122283

107.00 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 17463703

Quantity Price
1 113.00 € / Pack of 200
5 103.88 € / Pack of 200
Estimated Shipment: 30-12-2024
to see stock.
Explore more special offers
Add to basket

This item is not returnable. View return policy

Description

Description

  • Surface-coating-free, including substances like silicone, plasticizers, biocides or mold release agents to reduce the risk of sample interference.
  • Lot-certified PCR clean purity grade guarantees freedom from human DNA, DNase, RNase, and PCR inhibitors.
  • Available in tube, microplate, and deep-well plate formats for convenient upscaling of experiments.
  • Snap, screw, and SnapTec™ cap options available.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HDPE
25 mL
100/Bag, 2 Bags
200 Tubes
Not Autoclavable
Conical
Microcentrifuge Tube (PCR Clean, Protein LoBind)
PCR Clean (Non-sterile)
Colorless
Polypropylene
200 Tubes (4 Bags x 50 Tubes)
Non-sterile
PCR Clean
Screw Cap
17,000 x g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers