missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Protino™ Ni-NTA Agarose
Brand: Macherey-Nagel™ 745400.1
26.70 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Specifications
Up to 50 Settled Agarosemg/mL | |
Batch | |
4°C to 8°C | |
Agarose |
1 mL | |
6 % Beaded Agarose (Cross-Linked), Precharged With Ni2+ | |
100 mL | |
NTA (nitrilotriacetic acid) |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Warning
Hazard Category
- Flammable liquid Category 3
- Skin sensitiser Category 1
Hazard Statement
- H226-Flammable liquid and vapour.
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
- MIXTURE LIST-Contains : ethanol (diluted < 20 %) (denatured with1%IPA/1%MEK, acc.2016/1867/EU), resin suspension with Ni in
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction