missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Protino™ Ni-NTA Agarose
28.80 € - 1095.00 €
Specifications
Binding Capacity | Up to 50 Settled Agarosemg/mL |
---|---|
Binding Property | Batch |
Matrix | 6 % Beaded Agarose (Cross-Linked), Precharged With Ni2+ |
Content And Storage | 4°C to 8°C |
Quantity | 100 mL |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
12909934
|
Macherey-Nagel™
745400.1 |
1 mL |
28.80 €
1mL |
Estimated Shipment: 22-01-2025
Log in to see stock. |
|||||
12718702
|
Macherey-Nagel™
745400.100 |
100 mL |
1095.00 €
100mL |
In Stock
Log in to see stock. |
|||||
Specifications
Up to 50 Settled Agarosemg/mL | |
6 % Beaded Agarose (Cross-Linked), Precharged With Ni2+ | |
100 mL | |
NTA (nitrilotriacetic acid) |
Batch | |
4°C to 8°C | |
Agarose |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Warning
Hazard Category
- Flammable liquid Category 3
- Skin sensitiser Category 1
Hazard Statement
- H226-Flammable liquid and vapour.
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
- MIXTURE LIST-Contains : ethanol (diluted < 20 %) (denatured with1%IPA/1%MEK, acc.2016/1867/EU), resin suspension with Ni in
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title