missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ PRP-h5 Bulk Resin

Reversed phase mode.

Brand:  Hamilton™ 79280

Product Code. 12650343

  • 118.00 € / Each
Estimated Shipment: 06-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Particle size of 12 to 20μm
  • PSDVB packing material
  • Restoration: run a gradient of 100% water to 100% acetonitrile (repeat three times)
  • Stationary phase: PRP-h5

Specifications

Specifications

12 to 20 μm
Run a gradient of 100% water to 100% acetonitrile (repeat three times)
PRP-h5 columns
300 Å
Reversed phase
Bulk Resin
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.