Learn More
Thermo Scientific™ PT100 External Probe
For Use With: Cimarec and Supernuova Hotplates and Hotplate Stirrers
Brand:  Thermo Scientific™ CIC0001447
58.61 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Product Code. 15463660
  • 61.81 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
For Use With: Cimarec and Supernuova Hotplates and Hotplate Stirrers
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PT100 External probe, 0-400°C, Stainless steel, for all sizes of Cimarec+ and SuperNuova+ Hotplates and Hotplate Stirrers
0°C to 400°C
Digital
Cimarec and Supernuova Hotplates and Hotplate Stirrers
Probe
Stainless Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ PT100 External Probe > PT100 External probe, 0-400°C