missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ PTFE Dipper

Ideal for obtaining surface samples from a wide range of liquids. Sampling Systems™ PTFE Dippers are quick and easy to use.

Brand:  Sampling Systems™ 5100A-250

 View more versions of this product

Product Code. 15869361

  • 206.00 € / Each
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • PTFE is very durable and is resistant to chemical attack
  • PTFE product contact parts
  • Screw-in extension rods to increase the handle length
  • Suitable for use up to 280°C (400°F)
Specifications

Specifications

Dipper
Removable
Sampling Liquid
70 mm
PTFE
250 mL
Up to 280°C
600 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers