Learn More
Cowie Technology™ PTFE Joint Adapter
Creates high vacuum with sealing rings, negating the need for grease. Cowie Technology™ PTFE Joint Adapter are autoclavable and reusable.
Brand:  Cowie Technology™ 021.1014
 View more versions of this product
Product Code. 10591242
  • 9.75 € / Each
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Made from PTFE with a substantial shoulder for ease of handling
  • Overall robust construction for repeated use
  • Rigid sleeves and adapters are inert, usable to 280°C (approx.)
  • Withstands high vacuum
  • Transverse trapezoidal sealing rings on the contacting surfaces create high vacuum
Specifications
Specifications
14/23
PTFE Joint Adapters
1 Each
10/19