Learn More
Cowie Technology™ PTFE Joint Adapter
Creates high vacuum with sealing rings, negating the need for grease. Cowie Technology™ PTFE Joint Adapter are autoclavable and reusable.
9.75 € - 38.70 €
Specifications
Product Type PTFE Joint Adapters
Quantity 1 Each
View More Specs
Products 5
Product Code Brand Standard Taper Joint No. Cone Size Price Quantity & Availability  
Product Code Brand Standard Taper Joint No. Cone Size Price Quantity & Availability  
10782793
View Documents Promotion Details
Cowie Technology™
021.1924
19/26 29/32
14.50 €
Each
Estimated Shipment: 17-12-2024
to see stock.
Add to basket
10591242
View Documents Promotion Details
Cowie Technology™
021.1014
10/19 14/23
9.75 €
Each
Estimated Shipment: 26-11-2024
to see stock.
Add to basket
10531632
View Documents Promotion Details
Cowie Technology™
021.1424
10/19 24/29
14.50 €
Each
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
11393010
View Documents Promotion Details
Cowie Technology™
021.02945
29/42 45/50
32.45 €
Each
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
11303020
View Documents Promotion Details
Cowie Technology™
021.03445
35/45 45/50
38.70 €
Each
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
Description
Description
Made from PTFE with a substantial shoulder for ease of handling and an overall robust construction for repeated use. Rigid sleeves and adapters are inert, usable to 280°C (approx.) and will withstand high vacuum. High vacuum is obtained using transverse trapezoidal sealing rings on the contacting surfaces. For grease free, high vacuum seals using standard ground glass joints.
Specifications
Specifications
PTFE Joint Adapters
1 Each