Learn More
Bel-Art™ SP Scienceware™ PTFE Magnetic Stirring Bar Kit
Supplied in a convenient storage box and provide instant availability of PTFE encapsulated Alnico magnets.
Brand: Bel-Art™ F37161-0000
This item is not returnable.
View return policy
Description
- Useful for environmental testing and life science applications in which small sample volumes need to be prepared and evaluated
- Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon™ PTFE low friction coating; durable and inert
- Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
- Autoclave safe
Specifications
Autoclavable | |
25.4 x 8, 38.1 x 8, 50.8 x 8mm | |
Removable Pivot Rings | |
Alnico Magnet | |
Sphere |
ISO 9001:2008 | |
Vials, Micro test tubes, Centrifuge tubes | |
PTFE | |
1/Ea. | |
Magnetic Stir Bar |