missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ PTFE Magnetic Stirring Bar Kit
Supplied in a convenient storage box and provide instant availability of PTFE encapsulated Alnico magnets.
40.95 € - 273.00 €
Specifications
Autoclavable | Autoclavable |
---|---|
Quantity | 1/Ea. |
For Use With (Equipment) | Vials, Micro test tubes, Centrifuge tubes |
Product Code | Brand | Dimensions (L x W x H) | Shape | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W x H) | Shape | Price | Quantity & Availability | ||||
11735064
|
Bel-Art™
F37161-0000 |
25.4 x 8, 38.1 x 8, 50.8 x 8mm | Sphere |
64.82 €
Pack of 6 |
Estimated Shipment: 28-02-2025
Log in to see stock. |
||||
11725064
|
Bel-Art™
F37160-0000 |
25.4 x 8mm, 38.1 x 8mm, 50.8 x 8mm (Two Each) | Octagon |
40.95 €
Pack of 6 |
Estimated Shipment: 30-01-2025
Log in to see stock. |
||||
11745064
|
Bel-Art™
F37183-0000 |
12.7 x 7.92, 19.05 x 7.92, 25.4 x 7.92, 25.4 x 9.39, 38.1 x 7.92, 44.45 x 7.92, 50.8 x 9.39mm | Octagon |
273.00 €
Pack of 22 |
Estimated Shipment: 05-02-2025
Log in to see stock. |
||||
Description
- Useful for environmental testing and life science applications in which small sample volumes need to be prepared and evaluated
- Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon™ PTFE low friction coating; durable and inert
- Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
- Autoclave safe
Specifications
Autoclavable | |
1/Ea. | |
Vials, Micro test tubes, Centrifuge tubes |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title