Learn More
Cowie Technology™ PTFE Straight Through Stopcocks
Cowie Technology™ PTFE Straight Through Stopcocks feature a pure PTFE body and plug and combine all the desirable properties of the Ultimate Laboratory Stopcock.
Brand:  Cowie Technology™ 016.004
 View more versions of this product
Product Code. 10150192
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Medium vacuum pressure performance
  • Unbreakable
  • Chemically inert
  • Grease free
  • Supplied with borosilicate glass sidearms
Specifications
Specifications
Straight Through
Straight Through Stopcock
Autoclavable
1 Each
PTFE
4 mm
PTFE
10 mm
Borosilicate glass sidearms