Learn More
Cowie Technology™ PTFE Straight Through Stopcocks
Cowie Technology™ PTFE Straight Through Stopcocks feature a pure PTFE body and plug and combine all the desirable properties of the Ultimate Laboratory Stopcock.
Specifications
Type | Straight Through |
---|---|
Description | Straight Through Stopcock |
Material | PTFE |
Autoclavable | Autoclavable |
No. per Pack | 1 Each |
Description
- Medium vacuum pressure performance
- Unbreakable
- Chemically inert
- Grease free
- Supplied with borosilicate glass sidearms
Specifications
Straight Through | |
PTFE | |
1 Each | |
PTFE |
Straight Through Stopcock | |
Autoclavable | |
Borosilicate glass sidearms |