missing translation for 'onlineSavingsMsg'
Learn More

Cowie Technology™ PTFE Tall-form Evaporating Dish with Spout

Reduce spills when pouring excess liquid with integrated molded spout. Cowie Technology™ PTFE Tall-form Evaporating Dish with Spout includes a super-smooth surface for \maximum water and solvent evaporation.

Brand:  Cowie Technology™ 006.180

 View more versions of this product

Product Code. 10287900

  • 25.55 € / Each
Estimated Shipment: 03-01-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Isostatically molded in pure PTFE
  • Chemically resistant and inert
  • Super smooth internal finish
  • Usable to 280°C
  • Molded spout for easy pouring
Specifications

Specifications

11.7 fl oz.
180 mL
Round
Spout
-200°C to +280°C
80 mm
Flat
PTFE
Evaporating Dish
Smooth
50 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers