missing translation for 'onlineSavingsMsg'
Learn More

Cowie Technology™ PTFE Tubing

Cowie Technology™ PTFE Tubing is designed to be used with PTFE Bayonet Valves and Connectors.

Brand:  Cowie Technology™ 016.1706

 View more versions of this product

Product Code. 10722223

  • 14.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Chemically inert
  • Non-contaminating
  • Usable to 280°C
  • Manufactured from Virgin PTFE materials with FDA and USP Class VI approvals
Specifications

Specifications

Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
4 mm
6 mm
1 mm
-200°C
White
280°C
Autoclavable
PTFE
1 m
PTFE Bayonet Connectors & Valves
1 Each
Flexible
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.