missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ PTFE Watch Glass
PTFE Watch Glasses are ideally suited for covering beakers.
Brand: BRAND™ 150907
15.60 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Constructed of PTFE
- Ideal for covering PTFE beakers
- Excellent chemical resistance
Specifications
PTFE | |
Watch Glass | |
Beaker | |
65 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction