Learn More
Description
PTP1B Inhibitor is used as a PTP1B inhibitor blocks the activity of the Protein Tyrosine Phosphatase 1B, which is a key factor in the negative regulation of insulin and leptin signaling pathway. Protein tyrosine phosphatase 1B (PTP1B) has critical roles in cell signaling relevant to diabetes, obesity, and cancer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- PTP1B inhibitor blocks the activity of the Protein Tyrosine Phosphatase 1B, which is a key factor in the negative regulation of insulin and leptin signaling pathway

Specifications
Specifications
Quantity | 5 mg |
Description | White |
Content And Storage | -30°C to -10°C |
For Use With (Application) | PTP1B inhibitor blocks the activity of the Protein Tyrosine Phosphatase 1B, which is a key factor in the negative regulation of insulin and leptin signaling pathway |
CAS | 765317-72-4 |
Molecular Formula | C26H19Br2N3O7S3, C26H19Br2N3O7S3 |
Formula Weight | 741.45g/mol |
Synonym | 3-(3,5-Dibromo-4-hydroxybenzoyl)-2-ethyl-N-[4-(2-thiazolylsulfamoyl)phenyl]benzofuran-6-sulfonamide |
Sensitivity | Light Sensitive |
Solubility | Soluble in ethanol and DMSO |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
U19H013 | Certificate of Analysis | 07/12/2021 | J64512.#0 |
T11E012 | Certificate of Analysis | 07/12/2021 | J64512.#0 |
N04C009 | Certificate of Analysis | 07/12/2021 | J64512.#0 |
Z06C025 | Certificate of Analysis | 06/12/2021 | J64512.#0 |
W11A020 | Certificate of Analysis | 06/12/2021 | J64512.#0 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.