missing translation for 'onlineSavingsMsg'
Learn More

Labcon™ PurePlus™ 96-well PCR Plates for Roche™ Lightcycler™

These 96-well PCR plates come in natural color or opaque white.

Brand:  Labcon™ 3962-520-000-9

 View more versions of this product

Product Code. 17597946

  • 71.80 € / Pack of 10
Estimated Shipment: 12-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • ATP, RNase/DNase and Endotoxin-free (non-pyrogenic)
Specifications

Specifications

8 x 12
Polypropylene
96
10/Pk.
96.2 mm
Natural
0.1 mL
100/Cs.
128.6 mm
16.6 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers