missing translation for 'onlineSavingsMsg'
Learn More
HALYARD™ PUREZERO™ HG3 White Nitrile Gloves Product Code.: 17394320
Estimated Shipment: 29-04-2025
to see stock.
Add to basket

HALYARD™ PUREZERO™ HG3 White Nitrile Gloves
BIOPHARMAGREENER_CHOICE

Product Code. 17394320
X-Large
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
This item is not returnable. View return policy

Product Code. 17394320

Brand: HALYARD™ CLN3031XL

Estimated Shipment: 29-04-2025
to see stock.
Add to basket
Add to basket
This item is not returnable. View return policy

High-quality gloves designed to the exact standards to meet the requirements of a contamination controlled environment.

  • Low particulate counts and endotoxin levels due to repeated washes in deionized water and chlorination
  • Nickel-free
  • Powder-free
  • Silicone-free
  • Latex-free and no phtalates
  • High cleanliness
  • Low endotoxin levels
  • Formulated with ZERO presence of these hazardous traditional accelerators: Sulfur, Thiurams, Thiazoles, Guanidine, Carbamates(ZDEC or ZDBC)
Greener Choice Claims
  • Environmental benefits include:
    • Recycled Content
    • Renewable Energy
    • Source Reduction

Specifications

Certifications/Compliance ISO 3, ISO 4, ISO 5, ISO 6, ISO 7
Certifications/Compliance (Europe) GMP Grade A/B/C/D
Chemotherapeutic Rated ASTM D6978
Chlorinated Yes
Cleanroom Class Class 10 (ISO 4)
Accelerator Free Yes
AQL 1
Color White
Disposable Single-use
Exterior Finish Fully Textured
Finger Thickness (English) 6 mil
Finger Thickness (Metric) 0.16 mm
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Palm Thickness (English) 5 mil
Palm Thickness (Metric) 0.13 mm
Powdered Powder Free
Product Type Controlled Environment Gloves
Size X-Large
Sterility Non-sterile
Thickness (English) 6 mil
Thickness (Metric) 0.16 mm
Manufacturing/QMS Audit Standards ISO 9001:2015, EN13485:2016
Length (English) 12 in.
Length (Metric) 30.5 cm
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberSM429713X Certificate TypeCertificate of Analysis Date26/02/2025 Catalog NumberCLN3031XL
Lot NumberSM429723X Certificate TypeCertificate of Analysis Date13/02/2025 Catalog NumberCLN3031XL
Lot NumberSM416823X Certificate TypeCertificate of Analysis Date13/12/2024 Catalog NumberCLN3031XL
Lot NumberSM404123X Certificate TypeCertificate of Analysis Date01/08/2024 Catalog NumberCLN3031XL
Lot NumberSM302423X Certificate TypeCertificate of Analysis Date31/05/2023 Catalog NumberCLN3031XL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
HALYARD™ PUREZERO™ HG3 White Nitrile Gloves >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.