missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ PVC/PMP/FEP Graduated Burette

Constructed of a combination of plastics for good resistance to chemicals. Kartell™ PVC/PMP/FEP Graduated Burette offers a leakproof seal and eliminates the danger of glass breakages.

Brand:  Kartell™ 0199800

Product Code. 10632512

  • 121.00 € / Each
Estimated Shipment: 10-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • High precison plastic burettes
  • According to DIN 12.700 standards - Class B
  • Useful for routine work to eliminate the danger of glass breakages
  • Combination of different plastic materials (PMP, FEP, PVC)
  • The stopcock assembly is moulded in one piece, with FEP key
  • No lubricant needed
  • Provides a leakproof seal
Specifications

Specifications

25 mL
FEP, PMP, PVC
FEP, PMMA, PTFE
Class B
480 mm
0.1 mL
Graduated Burette
DIN 12700
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kartell™ PVC/PMP/FEP Graduated Burette > Capacity: 25mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.