missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente Pycnometers Acc. to Gay Lussac Adjusted

Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.

76.00 € - 122.00 €

Specifications

Material Borosilicate Glass (3.3)
View More Specs

Products 4
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
10400903
View Documents
Lenz Laborglasintrumente
05630210
5 mL
76.00 €
Each
Estimated Shipment: 29-07-2025
to see stock.
Add to basket
 
10778271
View Documents
Lenz Laborglasintrumente
05630228
50 mL
91.00 €
Each
Estimated Shipment: 29-07-2025
to see stock.
Add to basket
 
10385781
View Documents
Lenz Laborglasintrumente
05630222
25 mL
81.00 €
Each
Estimated Shipment: 30-05-2025
to see stock.
Add to basket
 
10335881
View Documents
Lenz Laborglasintrumente
05630237
100 mL
122.00 €
Each
Estimated Shipment: 29-07-2025
to see stock.
Add to basket
 
Description

Description

  • Gay Lussac pycnometers acc. to DIN ISO 3507
  • Stopper NS 10/19 with capillary
  • Stopper top ground and polished
  • Adjusted to In (with individual certificate)
  • Accuracy: 0.001 cm³
  • Material: borosilicate glass 3.3
  • Capcity shown is nominal capacity
Specifications

Specifications

Borosilicate Glass (3.3)
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lenz Laborglasintrumente Pycnometers Acc. to Gay Lussac Adjusted

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.