Learn More
Lenz Laborglasintrumente Pycnometers acc. To Gay Lussac Unadjusted
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05630010
 View more versions of this product
Product Code. 10797701
  • 33.00 € / Each
Estimated Shipment: 01-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Gay Lussac pycnometers acc. to DIN ISO 3507
  • Unadjusted, stopper NS 10/19
  • With capillary, stopper top ground and polished
  • Imprint of nominal capacity at the bottom
  • Material: borosilicate glass 3.3
Specifications
Specifications
5 mL
Borosilicate Glass (3.3)